Would you like to log in again?
Swingline™ GBC® ProClick® P110 Binding System
Supplier: Janitorial Supplies
A quick and affordable way to bind 11" documents. This manual system will bind ProClick spines fast to meet tight deadlines and still deliver a professionally finished look. Pages turn smoothly and lie flat. Exclusive new Guide Right paper guide aligns paper for accurate punching every time. No more misaligned punches! Easily add or delete pages without a machine. ProClick zip tool conveniently zips and unzips bound documents for quick edits. Recommended for small offices with monthly usage.
Ordering information: Manufacturer's limited one-year warranty.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Binding Spine Type Binding Sheet Capacity Punching Sheet Capacity Color Supplier No. VWR Catalog Number Unit Price Quantity
Comb 110 15 Gray/Silver SWI7708185 101389-230 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
W×D×H 18 x 14 x 10"
Max. Binding Edge 11"
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty