Human Recombinant NMNAT1 (highly active) (from E. coli)
Supplier: ProSci
90-354
75794-302EA 464.46 USD
75794-302
Human Recombinant NMNAT1 (highly active) (from E. coli)
Proteins and Peptides
Nuclear NMNAT isoform. Catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. It can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Interacts with PARP-1/ARTD1. Protects against axonal degeneration following mechanical or toxic insults. Widely expressed.
  • Fusion-Tag: His Tag
This recombinant proteins is for research use only.