Human Recombinant ANGPTL6 (from HEK293 cells)
Supplier: ProSci
90-019
75793-544EA 455.3 USD
75793-544
Human Recombinant ANGPTL6 (from HEK293 cells)
ANGPTL6 (Angiopoietin-like protein 6) regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism. The serum levels of ANGPTL6 are significantly increased in obese humans, patients with type 2 diabetes mellitus (T2DM), and in metabolic syndrome.
  • Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.