Nampt Recombinant Protein, Prosci
90-014
75793-534EA 588.12 USD
75793-534
Nampt Recombinant Protein, Prosci
Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
  • Fusion-Tag: His Tag
This recombinant proteins is for research use only.