Human Recombinant Complement Component C3a (from E. coli)
Supplier: ProSci
92-710
75791-982EA 664.62 USD
75791-982
Human Recombinant Complement Component C3a (from E. coli)
Complement is defined as key part of innate immunity and as the first line of defense in the fight against invading pathogens. Complement 3 (C3) is the most abundant component of the complement cascade and the convergent point for all three major complement activation pathways: namely classical, alternative and mannose-binding lectin pathways. Complement activation leads to the formation of the C3 convertase, which cleaves C3 into the key effector molecules, C3a (anaphylatoxin) and C3b (opsonin) which then drive microbe removal. By binding to C3a receptor (C3aR), C3a exhibits potent anaphylatoxin activity, including increased vascular permeability, triggering degranulation of mast cells, inflammation, and activating leukocytes.
This recombinant protein can be used for biological assays. For research use only.