Human Recombinant CD30L Receptor (from Cells)
Supplier: ProSci
92-672
75791-906EA 524.61 USD
75791-906
Human Recombinant CD30L Receptor (from Cells)
Proteins and Peptides
CD30, also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family, which regulates proliferation/apoptosis and antibody responses. CD30 is expressed by activated, but not by resting, T and B cells. Aberrant expression of CD30 by mastocytosis mast cells and interaction with its ligand CD30L (CD153) appears to play an important role in the pathogenesis and clinical presentation of systemic mastocytosis. CD30 has been considered as a specific diagnostic biomarker of anaplastic large cell lymphoma (ALCL) and classical Hodgkin lymphoma (cHL). CD30 is also a biomarker used for targeted therapy by an antibody–drug conjugate.
  • Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.