Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fc gamma RI Recombinant Protein, Prosci

Supplier: Prosci
92-647
CD64, also known as Fc-gamma receptor 1 (Fc gamma RI), is a type of integral membrane glycoprotein that binds monomeric IgG-type antibodies with high affinity. After binding IgG, CD64 interacts with an accessory chain known as the common gamma chain ( gamma chain), which possesses an ITAM motif that is necessary for triggering cellular activation. CD64 is composed of a signal peptide, three extracellular immunoglobulin domains of the C2-type used to bind antibody, a hydrophobic transmembrane domain, and a short cytoplasmic tail. CD64 mediates endocytosis, phagocytosis, antibody-dependent cellular cytotoxicity, cytokine release, and superoxide production. It is normally expressed on the surfaces of monocytes and macrophages.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg 92-647 75791-856 Each (50µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Fc gamma RI
Protein Synonyms FcRI, Fc-gamma RI, IgG Fc receptor I, FCGR1A, High affinity immunoglobulin gamma Fc receptor I, CD64
Protein/Peptide Type Recombinant
Species Mouse
Sequence Glu25-Pro297
Purity > 95% as determined by reducing SDS-PAGE
Endotoxin Content <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
Molecular Weight 31.5 kD
Formulation Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Storage Conditions Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Modification C-6 His tag

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty