Human Recombinant Cornulin (from E. coli)
Supplier: ProSci
91-100
75788-924EA 744.04 USD
75788-924
Human Recombinant Cornulin (from E. coli)
Proteins and Peptides
Cornulin is a member of the fused gene family of molecular chaperones. Human Cornulin contains N-terminus EF-hand domains and Ca2+ binding domains, and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. Cornulin involves in the mucosal/epithelial immune response and epidermal differentiation. Cornulin is a survival factor that participates in the clonogenicity of squamous esophageal epithelium cell lines, attenuates deoxycholic acid (DCA)-induced apoptotic cell death and release of calcium. When Cornulin is overexpressed in oral squamous carcinoma cell lines, it regulates negatively cell proliferation by the induction of G1 arrest.
  • Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.