Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CXCL4 Recombinant Protein, Prosci

Supplier: Prosci
91-044
Human Chemokine (C-X-C motif) Ligand 4 (CXCL4) is expressed in megakaryocytes and stored in the alpha-granules of platelets. CXCL4 contains several heparin-binding sites at the C-terminal region and binds heparin with high affinity. The active CXCL4 protein is a tetramer. Human and mouse CXCL4 share 64% sequence identity. CXCL4 is chemotactic for neutrophils, fibroblasts and monocytes and plays a critical role in inflammation and wound repair. CXCL4 functions via a splice variant of the chemokine receptor CXCR3, known as CXCR3B. The major physiologic role of CXCL4 appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. In contrast to other CXC chemokines, CXCL4 lacks chemotactic activity for polymorphonuclear granulocytes.

This recombinant protein can be used for biological assays. For research use only.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg 91-044 75788-812 Each (50µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name CXCL4
Protein Synonyms Oncostatin-A, Iroplact, PF4, Platelet Factor 4, C-X-C Motif Chemokine 4, PF-4, SCYB4, CXCL4
Protein/Peptide Type Recombinant
Species Human
Source E. coli
Sequence Glu32-Ser101
Purity > 95% as determined by reducing SDS-PAGE
Endotoxin Content <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
Molecular Weight 7.8 kD
Formulation Lyophilized from a 0.2 um filtered solution of 50mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Biological Activity ED50 is 5-15 μg/mL as determined by the ability of Recombinant CXCL4 to inhibit human FGF basic dependent proliferation of NR6R3T3 mouse fibroblasts.
Storage Conditions Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tested Applications Bioassay

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty