Anti-SERPINA12 Rabbit Polyclonal Antibody

Supplier: ProSci
36-251
75793-218EA 611.14 USD
75793-218
Anti-SERPINA12 Rabbit Polyclonal Antibody
Antibodies
Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a newtherapeutic target of metabolic syndrome.

ELISA: (direct and indirect: 1:2,000-1:5,000). Western Blot: (1:2,000-1:5,000 using ECL; suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk; suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: SERPINA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR