Anti-TNFSF11 Rabbit Polyclonal Antibody

Supplier: ProSci
36-214
75793-184EA 386.33 USD
75793-184
Anti-TNFSF11 Rabbit Polyclonal Antibody
RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T-cell proliferation. Important regulator of interactions between T-cells and dendritic cells and may play a role in the regulation of the T-cell-dependent immune response. Plays an important role in enhanced bone-resorption in humoral hypercalcemia of malignancy.

ELISA: (direct and indirect: 1:2,000-1:10,000). Flow Cytometry: (1:500-1:1,000). Western Blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is TBST with 0.05% Tween 20 with 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFSF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR