Anti-RETNLA Rabbit Polyclonal Antibody

Supplier: ProSci
36-204
75793-164EA 386.33 USD
75793-164
Anti-RETNLA Rabbit Polyclonal Antibody
RELM-alpha is a member to the family of adipocyte-secreted proteins called adipocytokines. It is abundantly expressed in white adipose tissue but not in preadipocytes, nor 3T3-L1 adipocytes. RELM-alpha is involved in the regulation of colonic inflammation and might be a link between colonic injury, glucose tolerance, and energy intake.

ELISA: (direct and indirect: 1:5,000-1:10,000). Western Blot: (1:5,000-1:10,000 using ECL. Suggested blocking and dilution buffer is TBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: Retnla
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR