Anti-LY6G Rat Monoclonal Antibody [clone: Nimp-R14]

Supplier: ProSci
36-155
75793-090EA 588.95 USD
75793-090
Anti-LY6G Rat Monoclonal Antibody [clone: Nimp-R14]
Antibodies
Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.

Flow Cytometry:(1:100). Immunohistochemistry:(1:200; frozen section / paraffin section) . Immunocytochemistry: (1:200). Functional (Blocking): 250ug / mouse (Optimal reagent to deplete neutrophils in vivo).

Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: Nimp-R14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR