Anti-FTO Mouse Monoclonal Antibody [clone: AG103]

Supplier: ProSci
36-093
75792-854EA 589.13 USD
75792-854
Anti-FTO Mouse Monoclonal Antibody [clone: AG103]
Antibodies
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

ELISA: (direct and indirect: 1:2,000-1:5,000). Immunoprecipitation: (5ug/ml per sample in a final reaction volume of 500-1,000ul). Immunohistochemistry: (5-10ug/ml: paraffin sections). Western Blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: Fto
Clonality: Monoclonal
Clone: AG103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR