Anti-DLK1 Mouse Monoclonal Antibody [clone: PF299-1]

Supplier: ProSci
36-071
75792-994EA 611.14 USD
75792-994
Anti-DLK1 Mouse Monoclonal Antibody [clone: PF299-1]
Antibodies
DLK1 (Protein delta homolog 1; Preadipocyte factor 1; Pref-1) is a regulator of adipocyte differentiation found in serum and urine. DLK1 might be a decoy ligand. Serum or plasma levels of soluble DLK1 affect negatively or positively adipogenesis and control mesenchymal cell fate. DLK1 is frequently upregulated in myelodysplastic syndrome (MDS) patients compared to non-leukemic individuals.

ELISA: (direct or indirect: 1:2,000-1:10,000). Flow Cytometry: (1:100-1:500). Immunohistochemistry: (paraffin sections (1:100-1:500)). Western blot: (1:2,000-1:10,000 using ECL. Suggested blocking and dilution buffer is TBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: PF299-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR