Anti-RETNLA Rat Monoclonal Antibody [clone: MREL 127]

Supplier: ProSci
36-061
75792-974EA 397.84 USD
75792-974
Anti-RETNLA Rat Monoclonal Antibody [clone: MREL 127]
RELM-alpha is a member to the family of adipocyte-secreted proteins called adipocytokines. It is abundantly expressed in white adipose tissue but not in preadipocytes, nor 3T3-L1 adipocytes. RELM-alpha is involved in the regulation of colonic inflammation and might be a link between colonic injury, glucose tolerance, and energy intake.

ELISA: (direct or indirect: 1:5,000-1:10,000). Western blot: (1:5,000-1:10,000 using ECL. Suggested blocking and dilution buffer is TBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: Retnla
Clonality: Monoclonal
Clone: MREL 127
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR