Anti-JAG1 Mouse Monoclonal Antibody [clone: J1G74-7]

36-053
75792-958EA 367.95 USD
75792-958
Anti-JAG1 Mouse Monoclonal Antibody [clone: J1G74-7]
Jagged-1 is a ligand for multiple Notch receptors and involved in the mediation of Notch signaling. It is involved in cell-fate decisions during hematopoiesis. Is involved in early and late stages of mammalian cardiovascular development. Inhibits myoblast differentiation. Enhances fibroblast growth factor-induced angiogenesis (in vitro). Defects in Jagged-1 are the cause of Alagille syndrome type 1 (ALGS1) and tetralogy of Fallot (TOF).

ELISA: (direct or indirect: 1:500-1:5,000). Flow Cytometry: (1:200-1:500) . Western blot: (1:500-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: J1G74-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR