Anti-SERPINA12 Mouse Monoclonal Antibody [clone: VP63]

Supplier: ProSci
36-047
75792-842EA 589.13 USD
75792-842
Anti-SERPINA12 Mouse Monoclonal Antibody [clone: VP63]
Antibodies
Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a newtherapeutic target of metabolic syndrome.

ELISA: (direct or indirect: 1:2,000-1:5,000). Western blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: SERPINA12
Clonality: Monoclonal
Clone: VP63
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR