Anti-ACE2 Mouse Monoclonal Antibody [clone: AC384]

Supplier: ProSci
36-038
75792-824EA 589.13 USD
75792-824
Anti-ACE2 Mouse Monoclonal Antibody [clone: AC384]
Antibodies
ACE2 has direct effects on cardiac functions. It is expressed predominantly in vascular endothelial cells of the heart and kidney. ACE2 converts angiotensin I to angiotensin 1-9. ACE2 was also identified as a functional receptor for SARS coronavirus.

ELISA: (direct or indirect: 1:2,000-1:10,000). Western blot: (1:2,000-1:10,000 using ECL. Suggested blocking and dilution buffer is PBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: AC384
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR