ESD Conductive Floor Runner, ACL Staticide
Supplier: ACL Staticide
6836 6848
10856-364EA 2313.23 USD
10856-364 10856-366
ESD Conductive Floor Runner, ACL Staticide
Floor Mats Permanent Floor Mats
The ACL Conductive Floor Runner is made from an engineered PVC with inherent electrical properties that will last the lifetime of the mat
ESD Conductive Floor Runners feature a single layer stippled surface. They are supplied in 75 foot lengths, with a choice between 36" and 48" widths. All floor runners are made from 100% recyclable material.
The homogeneous distribution of carbon black and proprietary polymers give this conductive thermoplastic vinyl a continuous and permanent path to ground. Resistance measures 10³ to 10⁶ ohms, per ANSI/ESD S7.1-2005.
Certifications: Compliant to ANSI/ESD S20.20.