Diammonium hydrogen citrate ≥98%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: Ammonium citrate dibasicDiammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylateCitric acid diammonium salt

AC61116-0025 AC61116-5000
200036-546EA 233.12 USD
200036-546 200005-736
Diammonium hydrogen citrate ≥98%
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Density: 1.483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

Specification Test Results

Appearance White Crystalline powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR