(±)-2-Butanol, extra dry 99+%, AcroSeal™

Warning

Synonyms: (±)-2-Hydroxy butane (±)-Ethyl methyl carbinol (±)-Methyl ethyl carbinol (±)-sec-Butyl alcohol (±)-sec-Butanol

AC61026-1000 AC61026-0010
103531-248EA 62.31 USD
103531-248 102775-128
(±)-2-Butanol, extra dry 99+%, AcroSeal™
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 99 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.81 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/03/13/710313.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR