Magnesium methyl carbonate 0.5 M in methanol, AcroSeal™

Danger

Synonyms: Methoxy(methyl carbonato-O)magnesiumMagnesium, methoxy(monomethyl carbonato-kappaO')-Methoxymagnesium methyl carbonate

AC42875-1000
102774-824EA 52.1 USD
102774-824
Magnesium methyl carbonate 0.5 M in methanol, AcroSeal™
Magnesium methyl carbonate

Appearance: Clear colorless Solution

Formula: C₃H₆MgO₄
MW: 130.382 g/mol
Density: 0.849 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008418
CAS Number: 4861-79-4
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Grignard 0.5 M Exact strength on the CoA
Titration Iodometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR