Tricyclohexylphosphine 1 M in toluene, AcroSeal™

Danger

AC42842-1000
102776-832EA 167 USD
102776-832
Tricyclohexylphosphine 1 M in toluene, AcroSeal™
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
MW: 280.43 g/mol
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.90 to 1.10 M
Melting point 81 to 84 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR