Ammonia 7N in methanol, AcroSeal™

Danger

AC42838-8000 AC42838-1000
102774-220EA 61.4 USD
102774-220 102776-140
Ammonia 7N in methanol, AcroSeal™
Ammonia
Formula: NH₃
MW: 17.03 g/mol
Boiling Pt: –33 °C (1013 hPa)
Melting Pt: –77 °C
Density: 0.77 g/cm³
Flash Pt: 14 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 6.0 to 7.5 N
Refractive index 1.6110 to 1.6130 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR