2,2-Dimethyl-4-pentenoic acid 97%

Danger

Synonyms: 2,2-Dimethylpent-4-enoic acid

AC42809-0010
102775-384EA 84.3 USD
102775-384
2,2-Dimethyl-4-pentenoic acid 97%
2,2-Dimethyl-4-pentenoic acid

Appearance: Clear colorless to yellow Liquid

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 104…108 °C (20 mmHg)
Density: 0.933 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00012304
CAS Number: 16386-93-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20 °C, 589 nm)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR