Ethyl-4-methyl-4-pentenoate 95%
|
Warning
|
Synonyms:
Ethyl 4-methylpent-4-enoate
AC42737-0010
AC42737-0050
102776-612EA
282
USD
102776-612
102774-954
Ethyl-4-methyl-4-pentenoate 95%
Ethyl-4-methyl-4-pentenoate
Appearance: Clear colorless Liquid
Formula:
C₈H₁₄O₂ MW: 142.198 g/mol Boiling Pt: 85 °C (20 mmHg) Density: 0.891 g/cm³ Flash Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075160 CAS Number: 4911-54-0 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...