Chloro(di-tert-butyl)phosphine 96%, AcroSeal™
|
Danger
|
Synonyms:
Di-t-butylchlorophosphine
, Di-tert-butylchlorophosphine
AC42728-0500
102777-402EA
358.62
USD
102777-402
Chloro(di-tert-butyl)phosphine 96%, AcroSeal™
Chloro(di-tert-butyl)phosphine
Formula:
C₈H₁₈ClP MW: 180.66 g/mol Boiling Pt: 69…70 °C (10 mmHg) Melting Pt: 2…3 °C Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008815 CAS Number: 13716-10-4 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥95 % |
UV | at 250 nm A: ≤0.01 |
UV | at 260 nm A: ≤0.005 |
UV | at 270 nm A: ≤0.005 |
UV | 1 cm cell vs HPLC-grade water |
UV | at 220 nm A: ≤0.2 |
UV | at 230 nm A: ≤0.1 |
UV | at 240 nm A: ≤0.05 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤2 ppm |
Acidity/alkalinity | ≤0.0001 meq/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...