Chloro(diisopropyl)phosphine 96%, AcroSeal™

Danger

Synonyms: Di-i-propylchlorophosphine

AC42727-0500
102776-758EA 305 USD
102776-758
Chloro(diisopropyl)phosphine 96%, AcroSeal™
Chloro(diisopropyl)phosphine
Formula: C₆H₁₄ClP
MW: 152.6 g/mol
Boiling Pt: 69 °C (33 mmHg)
Flash Pt: 3 °C
Storage Temperature: Ambient
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20 °C, 589 nm)
GC ≥95 %
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR