tert-Butylamine 99%, AcroSeal™

Danger

Synonyms: 2-Aminoisobutane

AC42724-1000 AC42724-8000
102776-876EA 44 USD
102776-876 102774-750
tert-Butylamine 99%, AcroSeal™
tert-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –67 °C
Density: 0.69 g/cm³
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20 °C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR