Triisopropyl borate 98+%, AcroSeal™
|
Danger
|
Synonyms:
Tri-i-propylborate
AC42714-1000
AC42714-8000
102774-350EA
50.7
USD
102774-350
102776-256
Triisopropyl borate 98+%, AcroSeal™
Triisopropyl borate
Formula:
C₉H₂₁BO₃ MW: 188.07 g/mol Boiling Pt: 139 °C (1013 hPa) Melting Pt: –59 °C Density: 0.814 g/cm³ Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008872 CAS Number: 5419-55-6 UN: 2616 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3755 to 1.3775 (20 °C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...