Sodium methoxide 0.5 M in methanol, AcroSeal™

Danger

Synonyms: Sodium methylateSodium methanolate

AC42722-8000 AC42722-1000
102776-304EA 222 USD
102776-304 102774-944
Sodium methoxide 0.5 M in methanol, AcroSeal™
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: –98 °C
Density: 0.8 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.45 to 0.60 M
UV at 250 nm A: ≤0.01
UV at 260 nm A: ≤0.005
UV 1 cm cell vs HPLC-grade water
UV at 200 nm A: ≤0.3
UV at 210 nm A: ≤0.15
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.02
Residue after evaporation ≤2 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR