(3R)-(+)-3-(Dimethylamino)pyrrolidine 98%

Danger

Synonyms: (R)-N,N-Dimethylpyrrolidin-3-amine(R)-(+)-3-(Dimethylamino)pyrrolidine

AC42699-0050 AC42699-0010
102775-966EA 121 USD
102775-966 102777-560
(3R)-(+)-3-(Dimethylamino)pyrrolidine 98%
(3R)-(+)-3-(Dimethylamino)pyrrolidine

Appearance: Clear colorless to yellow Liquid

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 166 °C (760 mmHg)
Flash Pt: 51.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00191347
CAS Number: 132958-72-6
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +12° to +16° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR