Methanol-[D₄] 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Danger

Synonyms: Carbinol-D4Methyl alcohol-D4Methanol-[²H₄]Methyl-D3 alcohol-DWood alcohol-D4Methanol-d4Hydroxy methane-D4Tetradeuteromethanol

AC42695-1000
102777-654EA 992.31 USD
102777-654
Methanol-[D₄] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 %
D-Enrichment ≥99.75 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR