(3-Acryloxypropyl)trimethoxysilane 90% stabilized

Danger

Synonyms: 3-(Trimethoxysilyl)propyl acrylate

AC42660-0250
103533-162EA 236 USD
103533-162
(3-Acryloxypropyl)trimethoxysilane 90% stabilized
(3-Acryloxypropyl)trimethoxysilane
Formula: C₉H₁₈O₅Si
Boiling Pt: 95 °C (3 mmHg)
Density: 1.059 g/cm³
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054803
CAS Number: 4369-14-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20 °C, 589 nm)
GC ≥90 %
Stabilizer Stabilized with BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR