4-Chloro-2-fluoro-5-nitrotoluene 98%
|
Warning
|
AC42601-0050
102775-218EA
79
USD
102775-218
4-Chloro-2-fluoro-5-nitrotoluene 98%
4-Chloro-2-fluoro-5-nitrotoluene
Formula:
C₇H₅ClFNO₂ Melting Pt: 42…45 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134231 CAS Number: 18349-11-6 |
Specification Test Results
Appearance | Yellow to light brown Crystalline mass |
Infrared spectrum | Conforms |
Melting point | 42 to 45 °C |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...