2,4,6-Trimethoxypyrimidine 98%

Warning

AC42674-0010 AC42674-2500
102774-882EA 92.9 USD
102774-882 103512-628
2,4,6-Trimethoxypyrimidine 98%
2,4,6-Trimethoxypyrimidine
Formula: C₇H₁₀N₂O₃
Boiling Pt: 232 °C (760 mmHg)
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00829225
CAS Number: 13106-85-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 51 to 54 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR