Methyl-4-trifluoromethylbenzoylacetate 95%

Synonyms: Methyl 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionoate4-(Trifluoromethyl)benzoylacetic acid methyll esterMethyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate3-Oxo-3-[4-(trifluoromethyl)phenyl]propionic acid methyll esterMethyl 4-(Trifluoromethyl)benzoylacetate

AC42544-0025
200013-258EA 82.1 USD
200013-258
Methyl-4-trifluoromethylbenzoylacetate 95%
Methyl-4-trifluoromethylbenzoylacetate

Appearance: Yellow Crystalline powder

Formula: C₁₁H₉F₃O₃
MW: 246.185 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00216521
CAS Number: 212755-76-5

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 62 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR