2-(4-Pyridyl)malondialdehyde 95%

Warning

Synonyms: 2-(Pyridin-4-yl)malonaldehyde

AC42534-0025
200008-816EA 146 USD
200008-816
2-(4-Pyridyl)malondialdehyde 95%
2-(4-Pyridyl)malondialdehyde

Appearance: Beige to brown Powder

Formula: C₈H₇NO₂
MW: 149.149 g/mol
Melting Pt: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00216535
CAS Number: 51076-46-1

Order Now

Specification Test Results

Appearance Beige to brown Powder
Infrared spectrum Conforms
Assay ≥94 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR