4-(Methylsulfonyl)phenol 95%

Warning

AC42488-0025
200004-332EA 92.5 USD
200004-332
4-(Methylsulfonyl)phenol 95%
4-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
MW: 172.20 g/mol
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00047806
CAS Number: 14763-60-1

Order Now

Specification Test Results

Appearance Tan to pink-brown Powder
Infrared spectrum Conforms
Melting point 90 to 95 °C
Water ≤5.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR