2-(Methylsulfonyl)benzoic acid 95%

Warning

AC42483-0025
200009-536EA 90.1 USD
200009-536
2-(Methylsulfonyl)benzoic acid 95%
2-(Methylsulphonyl)benzoic acid
Formula: C₈H₈O₄S
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00216492
CAS Number: 33963-55-2

Order Now

Specification Test Results

Appearance White to light yellow to light green Powder and/or chunks
Infrared spectrum Conforms
Melting point 136 to 141 °C
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR