Methyl-4-acetylbenzoate 99+%

Synonyms: Methyl 4-acetylbenzoate

AC42477-0050 AC42477-0010
200013-240EA 32.7 USD
200013-240 103514-940
Methyl-4-acetylbenzoate 99+%
Methyl 4-acetylbenzoate
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00216474
CAS Number: 3609-53-8

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 93 to 95 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR