Sodium nitroprusside dihydrate ACS, meets analytical specifications of Reagent Ph. Eur.

Danger

Synonyms: Sodium nitroferricyanide(III) dihydrateSodium nitrosopentacyanoferrate (III) dihydrateSodium nitroferricyanide dihydrateSodium pentacyanonitrosylferrate(III) dihydrateDisodium pentacyanonitrosylferrate (III) dihydrate

AC42436-5000
103528-756EA 354 USD
103528-756
Sodium nitroprusside dihydrate ACS, meets analytical specifications of Reagent Ph. Eur.
Sodium nitroprusside dihydrate
Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 12,08794

Order Now

Specification Test Results

Appearance Bordeaux Crystals
Titration Argentometric 99.0 to 102.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) Passes test (Limit about 0.01%)
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR