2-Chloroethylammonium chloride 98%

Warning

Synonyms: 2-Chloroethylamine hydrochloride2-Chloroethanamine hydrochloride

AC42286-0250 AC42286-1000 AC42286-0010
200005-026EA 57.8 USD
200005-026 103464-530 103464-532
2-Chloroethylammonium chloride 98%
2-Chloroethylammonium chloride
Formula: C₂H₇Cl₂N
MW: 115.99 g/mol
Melting Pt: 140…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00012887
CAS Number: 870-24-6
EINECS: 212-793-2
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder, crystals or chunks
Infrared spectrum Conforms
Melting point 140 to 150 °C
Loss on drying ≤1 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR