Vinylbenzyl trimethylammonium chloride 97%

Warning

Synonyms: Flocculant VA-2Ammonium, trimethyl(p-vinylbenzyl)-, chloride, polymers (8CI)4-Ethenyl-N,N,N-trimethylbenzenemethanaminium chloride homopolymer

AC42256-0250 AC42256-5000
103517-980EA 705 USD
103517-980 200015-152
Vinylbenzyl trimethylammonium chloride 97%
Vinylbenzyl trimethylammonium chloride
Formula: C₁₂H₁₈ClN
MW: 211.733 g/mol
Melting Pt: 240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080690
CAS Number: 26616-35-3

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR