Trimethylphenylammonium hydroxide 0.1 M in methanol

Danger

Synonyms: Trimethylanilinium HydroxideTMAHPhenyltrimethylammonium HydroxidePTAH

AC42202-1000 AC42202-5000 AC42202-0025
200058-802EA 506 USD
200058-802 200058-800 103528-686
Trimethylphenylammonium hydroxide 0.1 M in methanol
Trimethylphenylammonium hydroxide
Formula: C₉H₁₅NO
MW: 153.23 g/mol
Density: 0.8 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00041899
CAS Number: 1899-02-1
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 0.09 to 0.11 M
Melting point 69 to 73 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR