1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline) 99%

Warning

AC42192-0050
200033-710EA 420 USD
200033-710
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline) 99%
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline)

Appearance: Light brown to purple Powder

Formula: C₁₉H₁₉NO₂
Melting Pt: 149…150 °C
Storage Temperature: Ambient
CAS Number: 23001-29-8

Order Now

Specification Test Results

Appearance Light brown to purple Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR