D-ɑ-Tocopherol acetate 97%

Synonyms: Vitamin E Acetate (D-form)(+)-ɑ-Tocopherol Acetate

AC42106-0010
200018-688EA 43.3 USD
200018-688
D-ɑ-Tocopherol acetate 97%
D-alpha-Tocopherol acetate

Appearance: Clear yellow Viscous liquid

Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072052
CAS Number: 58-95-7

Order Now

Specification Test Results

Appearance Clear yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4990 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR