2-Isopropyl-N,2,3-trimethylbutyramide

Warning

Synonyms: N,2,3-Trimethyl-2-isopropylbutamide2-Isopropyl-N,2,3-trimethylbutanamide

AC42193-0050
200019-900EA 27.14 USD
200019-900
2-Isopropyl-N,2,3-trimethylbutyramide
2-Isopropyl-N,2,3-trimethylbutyramide
Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00130070
CAS Number: 51115-67-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
Water ≤0.5 %
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR