Sulfuric acid fuming (20 - 30% free SO₃), Extra Pure

Danger

Synonyms: Oleum

AC41997-0250 AC41997-5000 AC41997-0025
102777-624EA 956.54 USD
102777-624 102775-388 102776-052
Sulfuric acid fuming (20 - 30% free SO₃), Extra Pure
Sulfuric acid fuming
Formula: H₂O₇S₂ (H₂SO₄ + SO₃)
Melting Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00137792
CAS Number: 8014-95-7
UN: 1831
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Assay 20 to 30 % (free SO3)
Residue after ignition ≤0.01 %
Ammonium (NH4) ≤30 ppm
Arsenic (As) ≤2 ppm
Iron (Fe) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR